ENA432538810 Enamine

[4-(aminomethyl)-2-fluorophenyl]methanamine

  • MDL number MFCD16669391
  • Cas Number : 1017264-58-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-194750

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-194750
Availability details for
Quantity
  • Availability
  • Add to Cart