ENA432538811 Enamine

6-fluoro-1-benzofuran-3-carboxylic acid

  • MDL number MFCD22544066
  • Cas Number : 1393561-25-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-194753

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-194753
Availability details for
Quantity
  • Availability
  • Add to Cart