ENA432538812 Enamine

5-(2,3-dihydro-1H-indole-1-carbonyl)pyridin-2-amine

  • MDL number MFCD17439760
  • Cas Number : 1466285-94-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-194754

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-194754
Availability details for
Quantity
  • Availability
  • Add to Cart