ENA432538820 Enamine

2-benzenesulfonamido-4-hydroxybutanoic acid

  • MDL number MFCD13287472

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-194765

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-194765
Availability details for
Quantity
  • Availability
  • Add to Cart