ENA432538822 Enamine

3-oxo-3-(quinolin-6-yl)propanenitrile

  • MDL number MFCD01570559
  • Cas Number : 249937-48-2
  • Molecular Weight : 196.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-oxo-3-(quinolin-6-yl)propanenitrile
Inchi Code 1S/C12H8N2O/c13-6-5-12(15)10-3-4-11-9(8-10)2-1-7-14-11/h1-4,7-8H,5H2
Inchi Key FEDPKBUOSHROCN-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-194768

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-194768
Availability details for
Quantity
  • Availability
  • Add to Cart