ENA432538824 Enamine

[1-(4-bromophenyl)ethyl](heptyl)amine hydrochloride

  • MDL number MFCD28139559
  • Cas Number : 1788613-51-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-194770

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-194770
Availability details for
Quantity
  • Availability
  • Add to Cart