ENA432538826 Enamine

2,2,2-trifluoro-1-[1-(propan-2-yl)-1H-pyrazol-4-yl]ethan-1-one

  • MDL number MFCD18879665
  • Cas Number : 1342725-36-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-194772

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-194772
Availability details for
Quantity
  • Availability
  • Add to Cart