ENA432538834 Enamine

tert-butyl N-[2-(propan-2-yl)-1,3-thiazol-5-yl]carbamate

  • MDL number MFCD28139561
  • Cas Number : 1795467-03-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-194797

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-194797
Availability details for
Quantity
  • Availability
  • Add to Cart