ENA432538837 Enamine

1-[4-(aminomethyl)phenyl]-3,3-dimethylurea

  • MDL number MFCD11132968
  • Cas Number : 164648-47-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-194844

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-194844
Availability details for
Quantity
  • Availability
  • Add to Cart