ENA432538839 Enamine

4-chloro-1-methyl-1H-pyrazole-5-sulfonyl chloride

  • MDL number MFCD15976272
  • Cas Number : 1245807-31-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-194848

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-194848
Availability details for
Quantity
  • Availability
  • Add to Cart