ENA432539091 Enamine

4-(3-chlorophenyl)-4-methylpiperidine hydrochloride

  • MDL number MFCD28139564
  • Cas Number : 1803581-63-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-194855

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-194855
Availability details for
Quantity
  • Availability
  • Add to Cart