ENA432539095 Enamine

potassium 2-carbamoyl-2,2-difluoroacetate

  • MDL number MFCD28139571
  • Cas Number : 1803591-67-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data K+
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-194901

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-194901
Availability details for
Quantity
  • Availability
  • Add to Cart