ENA432539107 Enamine

(cyclobutylmethyl)(cyclopropylmethyl)amine hydrochloride

  • MDL number MFCD28139585
  • Cas Number : 1803572-40-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-195022

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195022
Availability details for
Quantity
  • Availability
  • Add to Cart