ENA432539108 Enamine

5-(hydrazinecarbonyl)-N,2,4-trimethyl-1H-pyrrole-3-carboxamide

  • MDL number MFCD28145257
  • Cas Number : 1803588-39-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-195031

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195031
Availability details for
Quantity
  • Availability
  • Add to Cart