ENA432539109 Enamine

3-[(2-methyl-3-phenylprop-2-en-1-yl)amino]propane-1,2-diol

  • Cas Number : 1807938-64-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-195035

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195035
Availability details for
Quantity
  • Availability
  • Add to Cart