ENA432539119 Enamine

1,1,1-trifluoro-3-methanesulfonylpropan-2-one

  • MDL number MFCD28145262
  • Cas Number : 1799297-94-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-195221

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195221
Availability details for
Quantity
  • Availability
  • Add to Cart