ENA432539123 Enamine

3-{[(4-bromo-5-methoxythiophen-2-yl)methyl]amino}propane-1,2-diol

  • MDL number MFCD28139609
  • Cas Number : 1803610-93-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-195265

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195265
Availability details for
Quantity
  • Availability
  • Add to Cart