ENA432539124 Enamine

ethyl 3-(methoxymethyl)piperidine-3-carboxylate

  • MDL number MFCD28145264
  • Cas Number : 1803582-67-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-195305

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195305
Availability details for
Quantity
  • Availability
  • Add to Cart