ENA432539127 Enamine

3-ethyl-1-methyl-1,4-diazepane dihydrochloride

  • MDL number MFCD28139615
  • Cas Number : 1803588-60-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-195431

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195431
Availability details for
Quantity
  • Availability
  • Add to Cart