ENA432539129 Enamine

(2,6-diethylphenyl)methanamine hydrochloride

  • MDL number MFCD28139618
  • Cas Number : 107694-43-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 240-243
Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-195466

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195466
Availability details for
Quantity
  • Availability
  • Add to Cart