ENA432539144 Enamine

N-(2-aminoethyl)-4-fluoro-N-methylbenzene-1-sulfonamide hydrochloride

  • MDL number MFCD28139633
  • Cas Number : 1803605-57-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-195628

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195628
Availability details for
Quantity
  • Availability
  • Add to Cart