ENA432539160 Enamine

3-(pyrimidin-2-yloxy)propan-1-amine dihydrochloride

  • MDL number MFCD28145292
  • Cas Number : 1803601-23-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-195914

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195914
Availability details for
Quantity
  • Availability
  • Add to Cart