ENA432539161 Enamine

2-[(1-cyclohexylethyl)amino]acetic acid hydrochloride

  • MDL number MFCD28145295
  • Cas Number : 1803600-49-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-195959

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195959
Availability details for
Quantity
  • Availability
  • Add to Cart