ENA432539186 Enamine

ethyl 2-amino-2-(4-methanesulfonylphenyl)propanoate hydrochloride

  • MDL number MFCD28145335
  • Cas Number : 1803592-52-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-196288

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-196288
Availability details for
Quantity
  • Availability
  • Add to Cart