ENA432539188 Enamine

(1r,3r)-3-(3-chloro-4-fluorophenyl)cyclobutan-1-amine

  • MDL number MFCD28145339
  • Cas Number : 1807936-48-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-196300

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-196300
Availability details for
Quantity
  • Availability
  • Add to Cart