ENA432539200 Enamine

5-(aminomethyl)-3,3-dimethyl-2,3-dihydro-1H-indol-2-one hydrochloride

  • MDL number MFCD28145355
  • Cas Number : 1803583-02-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-196485

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-196485
Availability details for
Quantity
  • Availability
  • Add to Cart