ENA432539210 Enamine

2-chloro-N-methyl-4-(trifluoromethyl)aniline hydrochloride

  • MDL number MFCD28145366
  • Cas Number : 1803589-01-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-196615

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-196615
Availability details for
Quantity
  • Availability
  • Add to Cart