ENA432539214 Enamine

5-chloro-2-fluoro-4-nitrobenzene-1-sulfonamide

  • MDL number MFCD28145370
  • Cas Number : 1803611-58-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-196637

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-196637
Availability details for
Quantity
  • Availability
  • Add to Cart