ENA432539217 Enamine

(2S)-2-amino-1-(pyrrolidin-1-yl)propan-1-one hydrochloride

  • MDL number MFCD28145378
  • Cas Number : 1807938-10-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-196690

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-196690
Availability details for
Quantity
  • Availability
  • Add to Cart