ENA432539225 Enamine

(1r,3r)-3-[4-(trifluoromethyl)phenyl]cyclobutan-1-amine hydrochloride

  • MDL number MFCD28145392
  • Cas Number : 1807939-34-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-196823

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
hcodes Xi
MSDS https://www.enaminestore.com/catalog/EN300-196823
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart