ENA432539228 Enamine

2-tert-butyl-1,3,4-thiadiazole

  • MDL number MFCD28145396
  • Cas Number : 317813-25-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-196881

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-196881
Availability details for
Quantity
  • Availability
  • Add to Cart