ENA432539295 Enamine

2-(methylamino)-N-[5-(2-methylpropyl)-1,3,4-thiadiazol-2-yl]acetamide dihydrochloride

  • MDL number MFCD28145480
  • Cas Number : 1803600-87-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-197042

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-197042
Availability details for
Quantity
  • Availability
  • Add to Cart