ENA432539296 Enamine

3-(ethylamino)pyrrolidin-2-one hydrochloride

  • MDL number MFCD28145481
  • Cas Number : 1803560-96-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-197053

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-197053
Availability details for
Quantity
  • Availability
  • Add to Cart