ENA432539303 Enamine

[5-(benzyloxy)-6-methoxypyridin-2-yl]methanamine

  • MDL number MFCD28145491
  • Cas Number : 1803562-77-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-197138

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-197138
Availability details for
Quantity
  • Availability
  • Add to Cart