ENA432539305 Enamine

3-bromo-1lambda4-thian-1-one

  • MDL number MFCD28145494
  • Cas Number : 332041-59-5
  • Molecular Weight : 197.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-bromotetrahydro-2H-thiopyran 1-oxide
Inchi Code 1S/C5H9BrOS/c6-5-2-1-3-8(7)4-5/h5H,1-4H2
Inchi Key WQMKSTDVFKPDJB-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-197167

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-197167
Availability details for
Quantity
  • Availability
  • Add to Cart