ENA432539310 Enamine

4-(difluoromethoxy)-3-(4-methoxyphenyl)aniline hydrochloride

  • MDL number MFCD28145501
  • Cas Number : 1803562-90-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-197312

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-197312
Availability details for
Quantity
  • Availability
  • Add to Cart