ENA432539311 Enamine

(2S,3S)-3-amino-1,1,1-trifluoro-4-methylpentan-2-ol

  • MDL number MFCD28145502
  • Cas Number : 291778-50-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 79-80
Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-197320

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-197320
Availability details for
Quantity
  • Availability
  • Add to Cart