ENA432539315 Enamine

4-bromo-3-chloro-N-[(1H-imidazol-2-yl)methyl]aniline

  • MDL number MFCD28145507
  • Cas Number : 1691144-33-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-197461

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-197461
Availability details for
Quantity
  • Availability
  • Add to Cart