ENA432539316 Enamine

4-bromo-3-chloro-N-[(pyridin-3-yl)methyl]aniline hydrochloride

  • MDL number MFCD28145508
  • Cas Number : 1803598-87-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-197462

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-197462
Availability details for
Quantity
  • Availability
  • Add to Cart