ENA432539318 Enamine

1-(cyclobutylmethyl)-1,4-diazepane dihydrochloride

  • MDL number MFCD28145512
  • Cas Number : 1803601-38-8
  • Molecular Weight : 241.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(cyclobutylmethyl)-1,4-diazepane dihydrochloride
Inchi Code 1S/C10H20N2.2ClH/c1-3-10(4-1)9-12-7-2-5-11-6-8-12;;/h10-11H,1-9H2;2*1H
Inchi Key AVPJVCQYVYLXDS-UHFFFAOYSA-N

Description

Purity 95
Salt Data Cl
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-197468

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-197468
Availability details for
Quantity
  • Availability
  • Add to Cart