ENA432539321 Enamine

4-fluoro-4-(3-methoxyphenyl)piperidine hydrochloride

  • MDL number MFCD28145517
  • Cas Number : 1345610-60-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-197578

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-197578
Availability details for
Quantity
  • Availability
  • Add to Cart