ENA432539334 Enamine

1,3-diethyl 2-(3-bromophenyl)-4-hydroxy-4-methyl-6-oxocyclohexane-1,3-dicarboxylate

  • MDL number MFCD01328593
  • Cas Number : 1803584-78-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-197894

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-197894
Availability details for
Quantity
  • Availability
  • Add to Cart