ENA432539336 Enamine

5-(chloromethyl)-1-phenyl-1H-1,2,4-triazole

  • MDL number MFCD20633541
  • Cas Number : 114370-76-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-194980

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-194980
Availability details for
Quantity
  • Availability
  • Add to Cart