ENA432539338 Enamine

2-(phenylmethylidene)-2H,3H,5H,6H,7H,8H,9H-imidazo[1,2-a]azepin-3-one

  • Cas Number : 306736-85-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-195497

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195497
Availability details for
Quantity
  • Availability
  • Add to Cart