ENA432539343 Enamine

3-methoxypyridine-2,6-dicarbonitrile

  • MDL number MFCD28139581
  • Cas Number : 1795466-51-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-194993

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-194993
Availability details for
Quantity
  • Availability
  • Add to Cart