ENA432539346 Enamine

1-(3-aminophenyl)-1H-imidazole-2-thiol hydrochloride

  • MDL number MFCD28139568
  • Cas Number : 1803572-32-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-194880

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-194880
Availability details for
Quantity
  • Availability
  • Add to Cart