ENA432539349 Enamine

1-(2,2-diethoxyethoxy)-4-(trifluoromethyl)benzene

  • MDL number MFCD18989309
  • Cas Number : 741290-08-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-195000

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195000
Availability details for
Quantity
  • Availability
  • Add to Cart