ENA432539350 Enamine

3-(2-methoxyethyl)-2,2-dimethyl-5-propylimidazolidin-4-one hydrochloride

  • MDL number MFCD28139628
  • Cas Number : 1786258-42-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-195517

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195517
Availability details for
Quantity
  • Availability
  • Add to Cart