ENA432539356 Enamine

1-(2-chloro-3-fluorophenyl)cyclopropan-1-ol

  • MDL number MFCD24111576
  • Cas Number : 1504931-98-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-195535

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195535
Availability details for
Quantity
  • Availability
  • Add to Cart