ENA432539359 Enamine

methyl 2-amino-5-cyclohexylthiophene-3-carboxylate

  • MDL number MFCD08059075
  • Cas Number : 1350623-54-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-195578

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195578
Availability details for
Quantity
  • Availability
  • Add to Cart